4,4'-Difluorbiphenyl ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
PFB Nr. 15, 1-Fluor-4-(4-fluorphenyl)benzol
L08361.03
L08361.06
L08361.03EA
31.6
EUR
L08361.03
L08361.06
4,4'-Difluorbiphenyl ≥99%
4,4'-Difluorbiphenyl
Purity: 99%
CAS No.: 398-23-2
Molecular Formula: C12H8F2
Formula Weight: 190.19
Melting Point: 87-89°
Boiling Point: 254-255°
Merck Reference: 14,3149
UN No.: 3152
MDL No.: MFCD00000349
Formel:
C₁₂H₈F₂ Molecular Weight: 190.19 g/mol Siedepunkt: 254…255 °C Schmelzpunkt: 87…89 °C Dichte: 1.361 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000349 CAS-Nummer: 398-23-2 EINECS: 206-910-6 ADR: 9 II Merck Index: 13,03173 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...