4-Fluorbiphenyl
Lieferant: FLUOROCHEM
Synonyme:
PFB Nr. 3, 1-Fluor-4-phenylbenzol
001670-1G
001670-5G
FLUO001670-1GEA
55.4
EUR
FLUO001670-1G
FLUO001670-5G
4-Fluorbiphenyl
4-Fluorbiphenyl
Formel:
C₁₂H₉F Molecular Weight: 172,2 g/mol Siedepunkt: 253 °C (1013 hPa) Schmelzpunkt: 59…61 °C Dichte: 1,247 g/cm³ (25 °C) |
MDL:
MFCD00011650 CAS-Nummer: 324-74-3 EINECS: 206-304-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...