2-Phenylanilin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Biphenyl-2-ylamin

L05728.03 L05728.06
L05728.03EA 18.8 EUR
L05728.03 L05728.06
2-Phenylanilin 97%
2-Phenylanilin

Purity: 97% CAS No.: 90-41-5 Molecular Formula: C12H11N Formula Weight: 169.23 Flash Point: 153°(307°F) Melting Point: 47-52° Boiling Point: 299° Beilstein Reference: 471874 MDL No.: MFCD00007701

Formel: C₁₂H₁₁N
Molecular Weight: 169,23 g/mol
Siedepunkt: 299 °C
Schmelzpunkt: 47…52 °C
Flammpunkt: 153 °C (307 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007701
CAS-Nummer: 90-41-5
EINECS: 201-990-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR