4,4'-Biphenol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Biphenyl-4,4'-diol, 4,4'-Dihydroxybiphenyl, 1-Hydroxy-4-(4-hydroxyphenyl)benzol

B22240.14 B22240.22 B22240.36
B22240.14EA 75.6 EUR
B22240.14 B22240.22 B22240.36
4,4'-Biphenol ≥99%
4,4'-Biphenol

Purity: 99% CAS No.: 92-88-6 Molecular Formula: C12H10O2 Formula Weight: 186.21 Odour: Odourless Melting Point: 279-285° Beilstein Reference: 1908886 MDL No.: MFCD00002348

Formel: C₁₂H₁₀O₂
Molecular Weight: 186.21 g/mol
Siedepunkt: 355
Schmelzpunkt: 279…285 °C
Dichte: 1.25 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002348
CAS-Nummer: 92-88-6
EINECS: 202-200-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR