Biphenyl (PCB Nr. 0) ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Diphenyl, PCB Nr. 0, 0-PCB, Phenylbenzol

A10265.0B A10265.0I A10265.30
A10265.0BEA 36 EUR
A10265.0B A10265.0I A10265.30
Biphenyl (PCB Nr. 0) ≥99%
Biphenyl (PCB Nr. 0)

Purity: 99% CAS No.: 92-52-4 Molecular Formula: C12H10 Formula Weight: 154.21 Flash Point: 113°(235°F) Odour: Pleasant Boiling Point: 254-255° Solubility: Soluble in alcohol and ether Beilstein Reference: 1634058 Merck Reference: 14,3314 UN No.: 3077 MDL No.: MFCD00003054

Formel: C₁₂H₁₀
Molecular Weight: 154,21 g/mol
Siedepunkt: 254…255 °C
Schmelzpunkt: 69…72 °C
Dichte: 0,990
Flammpunkt: 113 °C (235 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003054
CAS-Nummer: 92-52-4
EINECS: 202-163-5
UN: 3077
ADR: 9,III
Merck Index: 14,3314

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR