6-Chlor-4-chromanon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

640476-1G 640476-5G
SIAL640476-1GEA 57.7 EUR
SIAL640476-1G SIAL640476-5G
6-Chlor-4-chromanon, Sigma-Aldrich®
6-Chlor-4-chromanon
Formel: C₉H₇ClO₂
Molecular Weight: 182,61 g/mol
MDL: MFCD00024070
CAS-Nummer: 37674-72-9
EINECS: 253-611-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR