3-Hydroxycumarin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Hydroxy-2-benzopyron

642673-1G 642673-5G
SIAL642673-1GEA 42.7 EUR
SIAL642673-1G SIAL642673-5G
3-Hydroxycumarin, Sigma-Aldrich®
3-Hydroxycumarin
Formel: C₉H₆O₃
Molecular Weight: 162.14 g/mol
MDL: MFCD00017490
CAS-Nummer: 939-19-5
EINECS: 213-355-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR