4-Hydroxycumarin 98%
Lieferant: Thermo Scientific
Synonyme:
4-Hydroxy-2-benzopyron
121100050
121101000
121105000
ACRO121100050EA
20.5
EUR
ACRO121100050
ACRO121101000
ACRO121105000
4-Hydroxycumarin 98%
4-Hydroxycumarin
Formel:
C₉H₆O₃ Molecular Weight: 162,14 g/mol Siedepunkt: 349 °C (1013 hPa) Schmelzpunkt: 210…215 °C |
MDL:
MFCD00006856 CAS-Nummer: 1076-38-6 EINECS: 214-060-2 |
Spezifikation Testergebnisse
Appearance | White to yellow to beige Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 210 to 217 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...