4-Hydroxycumarin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Hydroxy-2-benzopyron
A14394.0E
A14394.22
A14394.36
A14394.0EEA
588
EUR
A14394.0E
A14394.22
A14394.36
4-Hydroxycumarin ≥98%
4-Hydroxycumarin
Purity: 98+%
CAS No.: 1076-38-6
Molecular Formula: C9H6O3
Formula Weight: 162.14
Odour: Odourless
Melting Point: ca 213° dec.
Beilstein Reference: 129768
MDL No.: MFCD00006856
Formel:
C₉H₆O₃ Molecular Weight: 162,14 g/mol Siedepunkt: 349 °C (1013 hPa) Schmelzpunkt: ∼ 213 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006856 CAS-Nummer: 1076-38-6 EINECS: 214-060-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...