6-Hydroxycumarin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Hydroxy-2-benzopyron

M-Clarity™ Qualitätsstufe = MQ100

642665-1G 642665-5G
SIAL642665-1GEA 44.4 EUR
SIAL642665-1G SIAL642665-5G
6-Hydroxycumarin, Sigma-Aldrich®
6-Hydroxycumarin
Formel: C₉H₆O₃
Molecular Weight: 162,14 g/mol
MDL: MFCD00630040
CAS-Nummer: 6093-68-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR