7-Amino-4-methylcumarin
Lieferant: BACHEM BIOCHEMICA
Synonyme:
7-Amino-4-methyl-2-benzopyron, AMC
4001606.0001
4001606.0005
BACE4001606.0001EA
0
EUR
BACE4001606.0001
BACE4001606.0005
7-Amino-4-methylcumarin
7-Amino-4-methylcumarin
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,19 g/mol Siedepunkt: 352 °C (1013 hPa) Schmelzpunkt: 226 °C Dichte: 1,377 g/cm³ (20 °C) |
MDL:
MFCD00006868 CAS-Nummer: 26093-31-2 EINECS: 247-454-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...