Benzo[a]pyren ≥96%
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: Benzo[def]chrysen, BaP
15856.02 15856.MC
15856.02EA 267 EUR
15856.02 15856.MC
Benzo[a]pyren ≥96%
Benzo[a]pyren
Purity: 96% CAS No.: 50-32-8 Molecular Formula: C20H12 Formula Weight: 252.30 Physical Form: Crystalline Appearance: Pale yellow/green/orange Melting Point: 177-180° Boiling Point: 495° Solubility: Soluble in benzene, toluene, and xylene. Sparingly soluble in alcohol, methanol. Insoluble in water Merck Reference: 14,1103 UN No.: 3077 MDL No.: MFCD00003602
Extensively used in cancer research
Formel: C₂₀H₁₂
Molecular Weight: 252.32 g/mol
Siedepunkt: 495 °C
Schmelzpunkt: 177…180 °C
Dichte: 1.351 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003602
CAS-Nummer: 50-32-8
EINECS: 200-028-5
UN: 3077
ADR: 9 III
Merck Index: 12,01134