2-Chinolincarbaldehyd, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-Chinolincarboxaldehyd

M-Clarity™ Qualitätsstufe = MQ100

334251-1G 334251-5G
SIAL334251-1GEA 68.7 EUR
SIAL334251-1G SIAL334251-5G
2-Chinolincarbaldehyd, Sigma-Aldrich®
2-Chinolincarbaldehyd
Formel: C₁₀H₇NO
Molecular Weight: 157.17 g/mol
MDL: MFCD00075032
CAS-Nummer: 5470-96-2
EINECS: 226-804-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR