2-Chlorlepidin ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Chlor-4-methylchinolin
H26504.06
H26504.14
H26504.06EA
55.9
EUR
H26504.06
H26504.14
2-Chlorlepidin ≥99%
2-Chlorlepidin
Purity: 99%
CAS No.: 634-47-9
Molecular Formula: C10H8ClN
Formula Weight: 177.63
Melting Point: 54-56°
Boiling Point: 296°
MDL No.: MFCD00006742
Formel:
C₁₀H₈ClN Molecular Weight: 177.63 g/mol Siedepunkt: 296 °C Schmelzpunkt: 54…56 °C Dichte: 1.23 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006742 CAS-Nummer: 634-47-9 EINECS: 211-209-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...