2,3-Dichlorchinoxalin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
A11734.06
A11734.14
A11734.06EA
46.3
EUR
A11734.06
A11734.14
2,3-Dichlorchinoxalin ≥98%
2,3-Dichloroquinoxaline
Purity: 98%
CAS No.: 2213-63-0
Molecular Formula: C8H4Cl2N2
Formula Weight: 199.04
Odour: Odourless
Melting Point: 151-155°
Beilstein Reference: 126076
UN No.: 2811
MDL No.: MFCD00006720
Formel:
C₈H₄Cl₂N₂ Molecular Weight: 199,04 g/mol Siedepunkt: 301 °C (1013 hPa) Schmelzpunkt: 151…155 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006720 CAS-Nummer: 2213-63-0 EINECS: 218-667-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...