6-Nitrochinoxalin

Lieferant: BIONET RESEARCH

12P-625.500 12P-625.1 12P-625.5
BIOT12P-625.500EA 0 EUR
BIOT12P-625.500 BIOT12P-625.1 BIOT12P-625.5
6-Nitrochinoxalin
6-Nitroquinoxaline
Formel: C₈H₅N₃O₂
Molecular Weight: 175.15 g/mol
MDL: MFCD00462822
CAS-Nummer: 6639-87-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR