Chinoxalin 98
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A14055.22
A14055.36
A14055.22EA
49.2
EUR
A14055.22
A14055.36
Chinoxalin 98
Quinoxaline
Purity: 98+%
CAS No.: 91-19-0
Molecular Formula: C8H6N2
Formula Weight: 130.15
Flash Point: 98°(208°F)
Melting Point: 27-32°
Boiling Point: 220-223°
Beilstein Reference: 109351
Merck Reference: 14,8078
MDL No.: MFCD00006719
Formel:
C₈H₆N₂ Molecular Weight: 130.15 g/mol Siedepunkt: 220…223 °C Schmelzpunkt: 27…32 °C Dichte: 1.124 Flammpunkt: 98 °C (208 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006719 CAS-Nummer: 91-19-0 EINECS: 202-047-4 Merck Index: 12,08263 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...