6-Methylchinoxalin

Lieferant: FLUOROCHEM
Achtung

002740-1G 002740-5G 002740-25G
FLUO002740-1GEA 31.1 EUR
FLUO002740-1G FLUO002740-5G FLUO002740-25G
6-Methylchinoxalin
6-Methylquinoxaline
Formel: C₉H₈N₂
Molecular Weight: 144.18 g/mol
MDL: MFCD00041001
CAS-Nummer: 6344-72-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR