Vitamin B12 (Cyanocobalamin), Millipore®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

1.24592.0100 1.24592.1000
1.24592.0100EA 43.8 EUR
1.24592.0100 1.24592.1000
Vitamin B12 (Cyanocobalamin), Millipore®
Vitamin B12 (Cyanocobalamin)
Formel: C₆₃H₈₈CoN₁₄O₁₄P
Molecular Weight: 1355,38 g/mol
Schmelzpunkt: 320 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00151092
CAS-Nummer: 68-19-9
EINECS: 200-680-0
Merck Index: 12,10152

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR