Tetramethyl-p-benzochinon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Tetramethyl-1,4-benzochinon, Tetramethylchinon, Durochinon, 2,3,5,6-Tetramethyl-1,4-benzochinon
D223204-1G
D223204-5G
SIALD223204-1GEA
78.3
EUR
SIALD223204-1G
SIALD223204-5G
Tetramethyl-p-benzochinon, Sigma-Aldrich®
Tetramethyl-p-benzochinon
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164.2 g/mol Siedepunkt: 282 Schmelzpunkt: 111…113 °C |
MDL:
MFCD00001604 CAS-Nummer: 527-17-3 EINECS: 208-409-8 Merck Index: 13,03503 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...