Cyclopentylisothiocyanat
Lieferant: FLUOROCHEM
|
Gefahr
|
002537-5G
002537-1G
FLUO002537-5GEA
198
EUR
FLUO002537-5G
FLUO002537-1G
Cyclopentylisothiocyanat
Cyclopentylisothiocyanat
Formel:
C₆H₉NS Molecular Weight: 127.21 g/mol Siedepunkt: 95…100 °C (1013 hPa) Dichte: 1.03 g/cm³ (20 °C) |
MDL:
MFCD00040877 CAS-Nummer: 33522-03-1 UN: 2922 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...