Ethinylcyclopentan 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
381310010
381310050
ACRO381310010EA
47.1
EUR
ACRO381310010
ACRO381310050
Ethinylcyclopentan 95%
Ethinylcyclopentan
Formel:
C₇H₁₀ Molecular Weight: 94.16 g/mol Flammpunkt: 1.1 °C |
MDL:
MFCD00013744 CAS-Nummer: 930-51-8 UN: 3295 |
Spezifikation Testergebnisse
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4470 to 1.4490 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...