Ethinylcyclopentan, Sigma-Aldrich®

Lieferant: Merck


MQ100

650331-1G 650331-5G
SIAL650331-1GEA 71.9 EUR
SIAL650331-1G SIAL650331-5G
Ethinylcyclopentan, Sigma-Aldrich®
Ethinylcyclopentan
Formel: C₇H₁₀
Molecular Weight: 94.16 g/mol
MDL: MFCD00013744
CAS-Nummer: 930-51-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR