1-Methylcyclopropancarbonsäure, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: 1-methylcyclopropane-1-carboxylic acid

205605-1G 205605-5G
SIAL205605-1GEA 78.9 EUR
SIAL205605-1G SIAL205605-5G
1-Methylcyclopropancarbonsäure, Sigma-Aldrich®
1-Methylcyclopropanecarboxylic acid
Formel: C₅H₈O₂
Molecular Weight: 100.12 g/mol
MDL: MFCD00001290
CAS-Nummer: 6914-76-7
EINECS: 230-020-7
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR