trans-4-Methylcyclohexancarbonsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
trans-4-Methyl-1-cyclohexanecarboxylic acid
H27455.03
H27455.06
H27455.03EA
44.4
EUR
H27455.03
H27455.06
trans-4-Methylcyclohexancarbonsäure ≥98%
trans-4-Methylcyclohexanecarboxylic acid
Purity: 98%
CAS No.: 13064-83-0
Molecular Formula: C8H14O2
Formula Weight: 142.20
Melting Point: 109-110°
MDL No.: MFCD00074943
Formel:
C₈H₁₄O₂ Molecular Weight: 142,2 g/mol Schmelzpunkt: 109…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00074943 CAS-Nummer: 13064-83-0 EINECS: 235-959-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...