Cyclopropylmethanol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Cyclopropylcarbinol, Cyclopropanmethanol
A12567.06
A12567.14
A12567.22
A12567.06EA
32.8
EUR
A12567.06
A12567.14
A12567.22
Cyclopropylmethanol ≥98%
Cyclopropylmethanol
Purity: 98%
CAS No.: 2516-33-8
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: 35°(95°F)
Refractive Index: 1.4330
Odour: Mild
Melting Point:
Formel:
C₄H₈O Molecular Weight: 72,11 g/mol Siedepunkt: 123…124 °C Schmelzpunkt: < –60 °C Dichte: 0,904 Flammpunkt: 35 °C (95 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001309 CAS-Nummer: 2516-33-8 EINECS: 219-735-5 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...