Cyclopropylmethanol
Lieferant: Thermo Scientific
Synonyme:
Cyclopropylcarbinol, Cyclopropanmethanol
SB00731.1
SB00731.10
SB00731.50
MAYBSB00731.1EA
0
EUR
MAYBSB00731.1
MAYBSB00731.10
MAYBSB00731.50
Cyclopropylmethanol
Cyclopropylmethanol
Formel:
C₄H₈O Molecular Weight: 72,11 g/mol Siedepunkt: 123 °C (1013 hPa) Schmelzpunkt: –60 °C Dichte: 0,904 g/cm³ (20 °C) |
MDL:
MFCD00001309 CAS-Nummer: 2516-33-8 EINECS: 219-735-5 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...