Bicyclohexyl, Sigma-Aldrich®

Lieferant: Merck


MQ200

D79403-25G D79403-100G D79403-5G
SIALD79403-25GEA 103 EUR
SIALD79403-25G SIALD79403-100G SIALD79403-5G
Bicyclohexyl, Sigma-Aldrich®
Bicyclohexyl
Formel: C₁₂H₂₂
Molecular Weight: 166.31 g/mol
Siedepunkt: 227
Dichte: 0.86 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003815
CAS-Nummer: 92-51-3
EINECS: 202-161-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR