2,2,4,4,6,6-Hexamethylcyclotrisilazan

Lieferant: CHEMPUR SILANE

Synonyme: 1,1,3,3,5,5-Hexamethylcyclotrisilazan

SILS09525.5 SILS09525.25 SILS09525.100
SILNSILS09525.5EA 0 EUR
SILNSILS09525.5 SILNSILS09525.25 SILNSILS09525.100
2,2,4,4,6,6-Hexamethylcyclotrisilazan
2,2,4,4,6,6-Hexamethylcyclotrisilazan
Formel: C₆H₂₁N₃Si₃
Molecular Weight: 219.51 g/mol
Siedepunkt: 186…188 °C (1013 hPa)
Dichte: 0.9196 g/cm³ (20 °C)
MDL: MFCD00039698
CAS-Nummer: 1009-93-4
EINECS: 213-773-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR