1-Ethylpiperazin ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N-Ethylpiperazine

L15539.36 L15539.14 L15539.22 AB173589.0025
L15539.36EA 159 EUR
L15539.36 L15539.14 L15539.22 ABCRAB173589.0025
1-Ethylpiperazin ≥98%
1-Ethylpiperazin

Purity: 98% CAS No.: 5308-25-8 Molecular Formula: C6H14N2 Formula Weight: 114.19 Flash Point: 43°(109°F) Refractive Index: 1.4690 Odour: Amine-like Boiling Point: 157° Beilstein Reference: 102971 UN No.: 2733 MDL No.: MFCD00059912

Formel: C₆H₁₄N₂
Molecular Weight: 114,19 g/mol
Siedepunkt: 157 °C
Schmelzpunkt: –60 °C
Dichte: 0,899
Flammpunkt: 43 °C (109 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00059912
CAS-Nummer: 5308-25-8
EINECS: 226-166-8
UN: 2733
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR