1-Phenylpiperazin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N-Phenylpiperazine
A12610.14
A12610.22
A12610.36
A12610.14EA
32.2
EUR
A12610.14
A12610.22
A12610.36
1-Phenylpiperazin ≥98%
1-Phenylpiperazin
Purity: 98+%
CAS No.: 92-54-6
Molecular Formula: C10H14N2
Formula Weight: 162.24
Flash Point: 141°(286°F)
Refractive Index: 1.5880
Melting Point: 17-19°
Boiling Point: 285-286°
Beilstein Reference: 132157
UN No.: 2927
MDL No.: MFCD00005957
Formel:
C₁₀H₁₄N₂ Molecular Weight: 162,23 g/mol Siedepunkt: 285…286 °C Schmelzpunkt: 17…19 °C Dichte: 1,064 Flammpunkt: 141 °C (286 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005957 CAS-Nummer: 92-54-6 EINECS: 202-165-6 UN: 2927 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...