2,7-Dibrom-9-fluorenon

Lieferant: Thermo Scientific

Synonyme: 2,7-Dibromo-9H-fluoren-9-one

BTB13291DA BTB13291EA
MAYBBTB13291DAEA 0 EUR
MAYBBTB13291DA MAYBBTB13291EA
2,7-Dibrom-9-fluorenon
2,7-Dibrom-9-fluorenon
Formel: C₁₃H₆Br₂O
Molecular Weight: 338 g/mol
MDL: MFCD00010790
CAS-Nummer: 14348-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR