1,8-Dichlor-9,10-anthrachinon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1,8-Dichloranthrachinon
M-Clarity™ Qualitätsstufe = MQ100
D56403-25G
D56403-100G
SIALD56403-25GEA
67.6
EUR
SIALD56403-25G
SIALD56403-100G
1,8-Dichlor-9,10-anthrachinon, Sigma-Aldrich®
1,8-Dichlor-9,10-anthrachinon
Formel:
C₁₄H₆Cl₂O₂ Molecular Weight: 277,11 g/mol |
MDL:
MFCD00001191 CAS-Nummer: 82-43-9 EINECS: 201-420-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...