1,5-Dichlor-9,10-anthrachinon 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1,5-Dichloroanthraquinone

A19534.18 A19534.30
A19534.18EA 44.1 EUR
A19534.18 A19534.30
1,5-Dichlor-9,10-anthrachinon 96%
1,5-Dichlor-9,10-anthrachinon

Purity: 96% CAS No.: 82-46-2 Molecular Formula: C14H6Cl2O2 Formula Weight: 277.11 Melting Point: 245-249° Beilstein Reference: 402592

Formel: C₁₄H₆Cl₂O₂
Molecular Weight: 277,11 g/mol
Siedepunkt: 401 °C (1013 hPa)
Schmelzpunkt: 245…249 °C
Dichte: 1,63 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001190
CAS-Nummer: 82-46-2
EINECS: 201-424-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR