Dibenzosuberon ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Dibenzosuberon, Dibenzo[b,f]cycloheptan-1-on, 1-Dibenzo[b,f]cycloheptanon
A18405.14
A18405.22
A18405.14EA
30.8
EUR
A18405.14
A18405.22
Dibenzosuberon ≥97%
Dibenzosuberon
Purity: 97%
CAS No.: 1210-35-1
Molecular Formula: C15H12O
Formula Weight: 208.26
Flash Point: 200°(392°F)
Refractive Index: 1.6340
Melting Point: 32-34°
Boiling Point: 147-149°/0.3mm
Beilstein Reference: 1454355
MDL No.: MFCD00003587
Formel:
C₁₅H₁₂O Molecular Weight: 208,26 g/mol Siedepunkt: 147…149 °C (0,3 mmHg) Schmelzpunkt: 32…34 °C Dichte: 1,156 Flammpunkt: 200 °C (392 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003587 CAS-Nummer: 1210-35-1 EINECS: 214-912-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...