9-Fluorenon ≥98%
Lieferant: Thermo Scientific
Synonyme:
9H-Fluoren-9-one
A12910.0E
A12910.22
A12910.36
A12910.0EEA
537
EUR
A12910.0E
A12910.22
A12910.36
9-Fluorenon ≥98%
9-Fluorenon
Purity: 98+%
CAS No.: 486-25-9
Molecular Formula: C13H8O
Formula Weight: 180.21
Flash Point: 163°(325°F)
Physical Form: Cryst powder or flakes
Density: 0.9
Appearance: Yellow
Melting Point: 82-85°
Boiling Point: 342°
Beilstein Reference: 1636531
MDL No.: MFCD00001141
Formel:
C₁₃H₈O Molecular Weight: 180,21 g/mol Siedepunkt: 342 °C Schmelzpunkt: 82…85 °C Dichte: 0,9 Flammpunkt: 163 °C (325 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001141 CAS-Nummer: 486-25-9 EINECS: 207-630-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...