1,5-Diamino-9,10-anthrachinon ≥90%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1,5-Diaminoanthrachinon
A18840.22
A18840.14
A18840.22EA
214
EUR
A18840.22
A18840.14
1,5-Diamino-9,10-anthrachinon ≥90%
1,5-Diamino-9,10-anthrachinon
Purity: 97%
CAS No.: 129-44-2
Molecular Formula: C14H10N2O2
Formula Weight: 238.25
Odour: Odourless
Melting Point: ca 320° dec.
MDL No.: MFCD00001226
Formel:
C₁₄H₁₀N₂O₂ Molecular Weight: 238,25 g/mol Schmelzpunkt: ∼ 320 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001226 CAS-Nummer: 129-44-2 EINECS: 204-947-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...