Dibenzofuran ≥98%
Lieferant: Thermo Scientific
Synonyme:
Diphenylenoxid, Dibenzo[b,d]furan
A16521.14
A16521.18
A16521.22
A16521.14EA
60.5
EUR
A16521.14
A16521.18
A16521.22
Dibenzofuran ≥98%
Dibenzofuran
Purity: 98%
CAS No.: 132-64-9
Molecular Formula: C12H8O
Formula Weight: 168.20
Flash Point: 130°(266°F)
Melting Point: 78-83°
Boiling Point: 287-289°
Beilstein Reference: 121100
UN No.: 3077
MDL No.: MFCD00004968
Formel:
C₁₂H₈O Molecular Weight: 168,19 g/mol Siedepunkt: 287…289 °C Schmelzpunkt: 78…83 °C Dichte: 1,3 Flammpunkt: 130 °C (266 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004968 CAS-Nummer: 132-64-9 EINECS: 205-071-3 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...