4-Brom-2-furaldehyd

Lieferant: FLUOROCHEM

011328-1G 011328-5G
FLUO011328-1GEA 121 EUR
FLUO011328-1G FLUO011328-5G
4-Brom-2-furaldehyd
4-Brom-2-furaldehyd
Formel: C₅H₃BrO₂
Lagertemperatur: Kühlschrank
MDL: MFCD00671504
CAS-Nummer: 21921-76-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR