4-Brom-2-furaldehyd, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Bromfurfural
M-Clarity™ Qualitätsstufe = MQ100
666599-1G
666599-250MG
SIAL666599-1GEA
71.8
EUR
SIAL666599-1G
SIAL666599-250MG
4-Brom-2-furaldehyd, Sigma-Aldrich®
4-Brom-2-furaldehyd
Formel:
C₅H₃BrO₂ Molecular Weight: 174,98 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00671504 CAS-Nummer: 21921-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...