5-Brom-2-furaldehyd 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Bromfurfural
329520100
329520010
ACRO329520100EA
97.8
EUR
ACRO329520100
ACRO329520010
5-Brom-2-furaldehyd 97%
5-Brom-2-furaldehyd
Formel:
C₅H₃BrO₂ Molecular Weight: 174,98 g/mol Siedepunkt: 201 °C (760 mmHg) Schmelzpunkt: 80…85 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00159501 CAS-Nummer: 1899-24-7 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to yellow to light brown Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 80 to 85 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...