5-Brom-2-furaldehyd
Lieferant: Thermo Scientific
XBX00321.1
XBX00321.10
XBX00321.50
MAYBXBX00321.1EA
0
EUR
MAYBXBX00321.1
MAYBXBX00321.10
MAYBXBX00321.50
5-Brom-2-furaldehyd
5-Brom-2-furaldehyd
Formel:
C₅H₃BrO₂ Siedepunkt: 201…203 °C (1013 hPa) Schmelzpunkt: 74…76 °C |
MDL:
MFCD00159501 CAS-Nummer: 1899-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...