5-Ethyl-2-furaldehyd ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Ethylfurfural

L06659.03 L06659.06
L06659.03EA 45.6 EUR
L06659.03 L06659.06
5-Ethyl-2-furaldehyd ≥98%
5-Ethyl-2-furaldehyd

Purity: 98% CAS No.: 23074-10-4 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 65°(149°F) Refractive Index: 1.5220 Boiling Point: 79-81°/12mm Beilstein Reference: 1982 MDL No.: MFCD00030605

Formel: C₇H₈O₂
Molecular Weight: 124,14 g/mol
Siedepunkt: 79…81 °C (12 mmHg)
Dichte: 1,055
Flammpunkt: 65 °C (149 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00030605
CAS-Nummer: 23074-10-4
EINECS: 245-411-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR