di-Natriumtetraborat, wasserfrei ≥99,95% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Borax, Sodium tetraborate
87862.14
87862.22
87862.14EA
73.4
EUR
87862.14
87862.22
di-Natriumtetraborat, wasserfrei ≥99,95% (Metall-Basis)
di-Natriumtetraborat
Purity: 99.95%
CAS No.: 1330-43-4
Molecular Formula: B4Na2O7
Formula Weight: 201.22
Physical Form: Powder
Density: 2.367
Refractive Index: 1.501
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 741°
Boiling Point: 1575° dec.
Merck Reference: 14,8590
MDL No.: MFCD00081185
Formel:
Na₂B₄O₇ Molecular Weight: 201,22 g/mol Siedepunkt: 1575 °C (Zers.) Schmelzpunkt: 741 °C Dichte: 2,367 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081185 CAS-Nummer: 1330-43-4 EINECS: 215-540-4 REACH: 01-2119490790-32 Merck Index: 14,8590 |