D(-)-Penicillamin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
H-D-Pen-OH, (S)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine
A11446.06
A11446.14
A11446.22
A11446.06EA
66.2
EUR
A11446.06
A11446.14
A11446.22
D(-)-Penicillamin 98%
D(-)-Penicillamin
Purity: 99%
CAS No.: 52-67-5
Molecular Formula: C5H11NO2S
Formula Weight: 149.21
Storage Temperature: 2°C to 8°C
Odour: Unpleasant
pH: 4.5-5.5
Melting Point: 202-204° dec.
Beilstein Reference: 1722375
Merck Reference: 14,7088
Optical Rotation: -63° (c=1 in 1N NaOH)
MDL No.: MFCD00064302
Formel:
C₅H₁₁NO₂S Molecular Weight: 149.2 g/mol Schmelzpunkt: 202…204 °C (Zers.) |
MDL:
MFCD00064302 CAS-Nummer: 52-67-5 EINECS: 200-148-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...