Natriumtetraethylborat 97%, rein

Lieferant: Thermo Scientific
Gefahr

ACRO363880050EA 370 EUR
ACRO363880050 ACRO363880010
Natriumtetraethylborat 97%, rein
Sodium tetraethylborate
Formel: C₈H₂₀BNa
Molecular Weight: 150,05 g/mol
Schmelzpunkt: 140…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061547
CAS-Nummer: 15523-24-7
UN: 3393
ADR: 4.2,

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR