Cäsiumcarbonat ≥99,9% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

10924.14 10924.06 10924.22 10924.36
10924.14EA 68.1 EUR
10924.14 10924.06 10924.22 10924.36
Cäsiumcarbonat ≥99,9% (Metall-Basis)
Cesium carbonate

Purity: 99.9% CAS No.: 534-17-8 Molecular Formula: Cs2CO3 Formula Weight: 325.82 Physical Form: -20 Mesh Powder/Granules Density: 4.072 pH: 10-12 Melting Point: 610° dec. Beilstein Reference: 4546405 Merck Reference: 14,2010 MDL No.: MFCD00010957

Formel: Cs₂CO₃
Molecular Weight: 325,82 g/mol
Schmelzpunkt: 610 °C (Zersetzung)
Dichte: 4,072
Lagertemperatur: Raumtemperatur
MDL: MFCD00010957
CAS-Nummer: 534-17-8
EINECS: 208-591-9
Merck Index: 14,2010

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR