Kaliumperiodat 99,8-100,3% ACS
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Kaliummetaperiodat, Kaliumtetroxoiodat(VII)
22896.14
22896.22
22896.14EA
64.9
EUR
22896.14
22896.22
Kaliumperiodat 99,8-100,3% ACS
Kaliumperiodat
Purity: 99.8%-100.3%
CAS No.: 7790-21-8
Molecular Formula: KIO4
Formula Weight: 230.00
Physical Form: Powder
Density: 3.618
Appearance: White
Melting Point: 582°
Merck Reference: 14,7654
UN No.: 1479
MDL No.: MFCD00011407
For the Colourimetric determination of Mn (oxidizes manganese compounds to permanganates), as oxidizing agent
Formel:
KlO₄ Molecular Weight: 230 g/mol Schmelzpunkt: 582 °C Dichte: 3,618 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011407 CAS-Nummer: 7790-21-8 EINECS: 232-196-0 UN: 1479 ADR: 5.1,II Merck Index: 14,7654 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...