Natriumhydroxid 98%, flake

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Caustic soda

A18395.0C A18395.0E A18395.36
A18395.0CEA 282 EUR
A18395.0C A18395.0E A18395.36
Natriumhydroxid 98%, flake
Sodium hydroxide

Purity: 98% CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Odour: Odourless pH: 14 Melting Point: 318° Boiling Point: 1390° Merck Reference: 14,8627 UN No.: 1823 MDL No.: MFCD00003548

Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 1390 °C
Schmelzpunkt: 318 °C
Dichte: 2.13
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8 II
REACH: 01-2119457892-27
Merck Index: 14,8627

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR