tetra-Natriumdiphosphat, wasserfrei, tech.
Lieferant: Thermo Scientific
Achtung
Synonyme: TSPP, tetra-Natriumpyrophosphat, Natriumpyrophosphat, Natriumpyrophosphat tetrabasisch, Natriumdiphosphat
13439.30 13439.A1 13439.99
13439.30EA 34.2 EUR
13439.30 13439.A1 13439.99
tetra-Natriumdiphosphat, wasserfrei, tech.
tetra-Natriumdiphosphat
CAS No.: 7722-88-5 Molecular Formula: Na4O7P2 Formula Weight: 265.90 Physical Form: Granular Density: 2.534 Sensitivity: Hygroscopic Appearance: White Melting Point: 988° Solubility: Soluble in water, with slow hydrolysis to sodium orthophosphate Merck Reference: 14,9240 MDL No.: MFCD00003513
Water softener, metal cleaner, soap and synthetic detergent builder, general sequestering agent, in electrodeposition of metals
Formel: Na₄O₇P₂
Molecular Weight: 265,9 g/mol
Schmelzpunkt: 988 °C
Dichte: 2,534
Lagertemperatur: Raumtemperatur
MDL: MFCD00003513
CAS-Nummer: 7722-88-5
EINECS: 231-767-1
Merck Index: 14,9240